3-Brombenzoylacetonitril

Lieferant: CHEMPUR

05-0047.1 05-0047.5
CHMP05-0047.1EA 0 EUR
CHMP05-0047.1 CHMP05-0047.5
3-Brombenzoylacetonitril
3-Brombenzoylacetonitril
Formel: C₉H₆BrNO
MDL: MFCD02260778
CAS-Nummer: 70591-86-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR