5-Brom-1H-indazol

Lieferant: CHEMPUR

45-0040.1 45-0040.5
CHMP45-0040.1EA 0 EUR
CHMP45-0040.1 CHMP45-0040.5
5-Brom-1H-indazol
5-Bromo-1H-indazole
Formel: C₇H₅BrN₂
Molecular Weight: 197,03 g/mol
MDL: MFCD00839493
CAS-Nummer: 53857-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR