1-(2-Fluor-4-nitrophenyl)piperazin

Lieferant: CHEMPUR

65-0001.1 65-0001.5
CHMP65-0001.1EA 0 EUR
CHMP65-0001.1 CHMP65-0001.5
1-(2-Fluor-4-nitrophenyl)piperazin
1-(2-Fluor-4-nitrophenyl)piperazin
Formel: C₁₀H₁₂FN₃O₂
Molecular Weight: 225.22 g/mol
MDL: MFCD01572463
CAS-Nummer: 154590-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR