1-Cyclohexylpiperazin

Lieferant: CHEMPUR

65-0022.1 65-0022.5
CHMP65-0022.1EA 0 EUR
CHMP65-0022.1 CHMP65-0022.5
1-Cyclohexylpiperazin
1-Cyclohexylpiperazin
Formel: C₁₀H₂₀N₂
Molecular Weight: 168.28 g/mol
Siedepunkt: 256
Schmelzpunkt: 28…32 °C
MDL: MFCD00044809
CAS-Nummer: 17766-28-8
EINECS: 241-750-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR