1-(3-Methylphenyl)piperazin
Lieferant: Thermo Scientific
AC13647.1
AC13647.10
AC13647.50
MAYBAC13647.1EA
0
EUR
MAYBAC13647.1
MAYBAC13647.10
MAYBAC13647.50
1-(3-Methylphenyl)piperazin
1-(3-Methylphenyl)piperazin
Formel:
C₁₁H₁₆N₂ Molecular Weight: 176,26 g/mol |
MDL:
MFCD00040734 CAS-Nummer: 41186-03-2 EINECS: 255-251-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...