5-(Trifluormethyl)-o-anisidin

Lieferant: Thermo Scientific

Synonyme: 3-Amino-4-methoxybenzotrifluorid, 5-(Trifluormethyl)-2-methoxyanilin, 2-Methoxy-5-(trifluormethyl)anilin, 2-Amino-4-(trifluormethyl)anisol, 5-(Trifluormethyl)-2-anisidin

BTB03143.1 BTB03143.10 BTB03143.50
MAYBBTB03143.1EA 0 EUR
MAYBBTB03143.1 MAYBBTB03143.10 MAYBBTB03143.50
5-(Trifluormethyl)-o-anisidin
5-(Trifluormethyl)-o-anisidin
Formel: C₈H₈F₃NO
Molecular Weight: 191.15 g/mol
Siedepunkt: 230
Schmelzpunkt: 59
MDL: MFCD00042486
CAS-Nummer: 349-65-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR