2-Nitro-1,3,5-trimethoxybenzol

Lieferant: Thermo Scientific

Synonyme: 1-Nitro-2,4,6-trimethoxybenzol, 2,4,6-Trimethoxynitrobenzol

BTB09099.1 BTB09099.10 BTB09099.50
MAYBBTB09099.1EA 0 EUR
MAYBBTB09099.1 CHMPBTB09099.10 CHMPBTB09099.50
2-Nitro-1,3,5-trimethoxybenzol
2-Nitro-1,3,5-trimethoxybenzol
Formel: C₉H₁₁NO₅
Molecular Weight: 213,19 g/mol
Schmelzpunkt: 153 °C
MDL: MFCD00016992
CAS-Nummer: 14227-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR