2-Amino-4,6-dimethoxypyrimidin
Lieferant: Thermo Scientific
Synonyme:
4,6-Dimethoxypyrimidin-2-amine, 4,6-Dimethoxypyrimidin-2-ylamine
BTB09643.1
BTB09643.10
BTB09643.50
MAYBBTB09643.1EA
0
EUR
MAYBBTB09643.1
MAYBBTB09643.10
MAYBBTB09643.50
2-Amino-4,6-dimethoxypyrimidin
2-Amino-4,6-dimethoxypyrimidine
Formel:
C₆H₉N₃O₂ Molecular Weight: 155,16 g/mol Siedepunkt: 276 °C (1013 hPa) Schmelzpunkt: 97 °C Dichte: 1,429 g/cm³ (–153 °C) |
MDL:
MFCD00038832 CAS-Nummer: 36315-01-2 EINECS: 252-969-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...