1-Brom-4-propoxybenzol

Lieferant: CHEMPUR

Synonyme: 4-Propoxybrombenzol

BTB09792.1 BTB09792.10 BTB09792.50
CHMPBTB09792.1EA 0 EUR
CHMPBTB09792.1 CHMPBTB09792.10 CHMPBTB09792.50
1-Brom-4-propoxybenzol
1-Bromo-4-propoxybenzene
Formel: C₉H₁₁BrO
Molecular Weight: 215,09 g/mol
MDL: MFCD00156160
CAS-Nummer: 39969-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR