3,5-Di-tert-butyl-4-hydroxybenzoesäure
Lieferant: CHEMPUR
BTB10318.250
BTB10318.1
CHMPBTB10318.250EA
0
EUR
CHMPBTB10318.250
MAYBBTB10318.1
3,5-Di-tert-butyl-4-hydroxybenzoesäure
3,5-Di-tert-butyl-4-hydroxybenzoesäure
Formel:
C₁₅H₂₂O₃ Siedepunkt: 374 °C (1013 hPa) Schmelzpunkt: 219…220 °C |
MDL:
MFCD00008827 CAS-Nummer: 1421-49-4 EINECS: 215-823-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...