5-Fluor-2-nitrobenzoesäure

Lieferant: CHEMPUR

BTB13065.250 BTB13065.1
CHMPBTB13065.250EA 0 EUR
CHMPBTB13065.250 MAYBBTB13065.1
5-Fluor-2-nitrobenzoesäure
5-Fluor-2-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
MDL: MFCD00055635
CAS-Nummer: 320-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR