5-Brom-2,4-dichlorpyrimidin
Lieferant: CHEMPUR
BTB14083.250
BTB14083.1
CHMPBTB14083.250EA
0
EUR
CHMPBTB14083.250
MAYBBTB14083.1
5-Brom-2,4-dichlorpyrimidin
5-Bromo-2,4-dichloropyrimidine
Formel:
C₄HBrCl₂N₂ Molecular Weight: 227.87 g/mol Siedepunkt: 288 Schmelzpunkt: –1 °C Dichte: 1.781 g/cm³ (20 °C) |
MDL:
MFCD00127818 CAS-Nummer: 36082-50-5 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...