5-Methyl-3-isoxazolcarbonsäure

Lieferant: Thermo Scientific

CC10401DA CC10401EA
MAYBCC10401DAEA 0 EUR
MAYBCC10401DA MAYBCC10401EA
5-Methyl-3-isoxazolcarbonsäure
5-Methylisoxazole-3-carboxylic acid
Formel: C₅H₅NO₃
Molecular Weight: 127.1 g/mol
MDL: MFCD01318162
CAS-Nummer: 3405-77-4
EINECS: 222-289-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR