2-Thenoesäure
Lieferant: Thermo Scientific
Synonyme:
Thiophen-2-carbonsäure, 2-Thiophencarbonsäure
CC13001.1
CC13001.10
MAYBCC13001.1EA
0
EUR
MAYBCC13001.1
MAYBCC13001.10
2-Thenoesäure
2-Thenoesäure
Formel:
C₅H₄O₂S Molecular Weight: 128,15 g/mol Siedepunkt: 260 °C (1013 hPa) Schmelzpunkt: 123…125 °C |
MDL:
MFCD00005437 CAS-Nummer: 527-72-0 EINECS: 208-423-4 Merck Index: 12,09491 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...