3-Thiophenmethanol

Lieferant: CHEMPUR

Synonyme: Thiophen-3-methanol, 3-Thienylmethanol, 3-(Hydroxymethyl)thiophen

CC13109.250 CC13109.1
CHMPCC13109.250EA 0 EUR
CHMPCC13109.250 MAYBCC13109.1
3-Thiophenmethanol
3-Thiophenmethanol
Formel: C₅H₆OS
Molecular Weight: 114.17 g/mol
MDL: MFCD00014534
CAS-Nummer: 71637-34-8
EINECS: 275-741-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR