1-(2-Nitrophenyl)pyrrol

Lieferant: Thermo Scientific

Synonyme: N-(2-Nitrophenyl)pyrrol

CD03294.1 CD03294.10 CD03294.50
MAYBCD03294.1EA 0 EUR
MAYBCD03294.1 MAYBCD03294.10 MAYBCD03294.50
1-(2-Nitrophenyl)pyrrol
1-(2-Nitrophenyl)pyrrole
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188,19 g/mol
MDL: MFCD00047062
CAS-Nummer: 33265-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR