1-(4-Chlorphenyl)pyrrol

Lieferant: Thermo Scientific

Synonyme: N-(4-Chlorphenyl)pyrrol

CD03747DA CD03747EA CD03747.50
MAYBCD03747DAEA 0 EUR
MAYBCD03747DA MAYBCD03747EA MAYBCD03747.50
1-(4-Chlorphenyl)pyrrol
1-(4-Chlorophenyl)pyrrole
Formel: C₁₀H₈ClN
Molecular Weight: 177,63 g/mol
Siedepunkt: 200…203 °C (95 mmHg)
Schmelzpunkt: 90 °C
MDL: MFCD00047067
CAS-Nummer: 5044-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR