2,6-Dimethoxybenzylalkohol

Lieferant: Thermo Scientific

CD04716.250 CD04716.1
MAYBCD04716.250EA 0 EUR
MAYBCD04716.250 MAYBCD04716.1
2,6-Dimethoxybenzylalkohol
2,6-Dimethoxybenzylalkohol
Formel: C₉H₁₂O₃
MDL: MFCD00016862
CAS-Nummer: 16700-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR