2-Chlor-6-fluorbenzylamin

Lieferant: CHEMPUR

CD05338.250 CD05338.1
CHMPCD05338.250EA 0 EUR
CHMPCD05338.250 MAYBCD05338.1
2-Chlor-6-fluorbenzylamin
2-Chlor-6-fluorbenzylamin
Formel: C₇H₇ClFN
Molecular Weight: 159,59 g/mol
Siedepunkt: 99…100 °C (1013 hPa)
MDL: MFCD00042458
CAS-Nummer: 15205-15-9
EINECS: 239-259-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR