2-Fluor-3-methylbenzylbromid

Lieferant: Thermo Scientific

Synonyme: 1-(Bromomethyl)-2-fluoro-3-methylbenzene

CD05588DA CD05588EA
MAYBCD05588DAEA 0 EUR
MAYBCD05588DA MAYBCD05588EA
2-Fluor-3-methylbenzylbromid
2-Fluor-3-methylbenzylbromid
Formel: C₈H₈BrF
Molecular Weight: 203,05 g/mol
Siedepunkt: 105…106 °C (10 mmHg)
MDL: MFCD00042470
CAS-Nummer: 151412-12-3
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR