α-Brom-3-chlor-2-fluortoluol
Lieferant: Thermo Scientific
Synonyme:
3-Chlor-2-fluorbenzylbromid
CD10824.1
CD10824.10
MAYBCD10824.1EA
0
EUR
MAYBCD10824.1
MAYBCD10824.10
α-Brom-3-chlor-2-fluortoluol
α-Brom-3-chlor-2-fluortoluol
Formel:
C₇H₅BrClF Molecular Weight: 223,47 g/mol Siedepunkt: 125…126 °C (15 mmHg) |
MDL:
MFCD00042503 CAS-Nummer: 85070-47-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...