1-(3-Brompropoxy)-4-nitrobenzol

Lieferant: Thermo Scientific

GK03637.1 GK03637.10
MAYBGK03637.1EA 0 EUR
MAYBGK03637.1 MAYBGK03637.10
1-(3-Brompropoxy)-4-nitrobenzol
1-(3-Bromopropoxy)-4-nitrobenzene
Formel: C₉H₁₀BrNO₃
Molecular Weight: 260,09 g/mol
Siedepunkt: 150…151 °C (0,2 mmHg)
Schmelzpunkt: 62…63 °C
CAS-Nummer: 13094-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR