N-(4-Chlorphenyl)phthalimid

Lieferant: CHEMPUR

Synonyme: N-(4-Chlorphenyl)-1,2-benzoldicarbonsäureimid, N-(4-Chlorphenyl)phthalsäureimid

IP33.100 IP33.250
CHMPIP33.100EA 0 EUR
CHMPIP33.100 CHMPIP33.250
N-(4-Chlorphenyl)phthalimid
N-(4-Chlorphenyl)phthalimid
Formel: C₁₄H₈ClNO₂
MDL: MFCD00023048
CAS-Nummer: 7386-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR