2'-Hydroxy-5'-methoxyacetophenon
Lieferant: Thermo Scientific
Synonyme:
2'-Hydroxy-5'-Methoxyacetophenone, 2′-Hydroxy-5′-methoxyacetophenone
JFD00869.1
JFD00869EA
JFD00869EE
MAYBJFD00869.1EA
0
EUR
MAYBJFD00869.1
MAYBJFD00869EA
MAYBJFD00869EE
2'-Hydroxy-5'-methoxyacetophenon
2'-Hydroxy-5'-methoxyacetophenon
Formel:
C₉H₁₀O₃ Molecular Weight: 166.18 g/mol Siedepunkt: 281 Schmelzpunkt: 113 |
MDL:
MFCD00008731 CAS-Nummer: 705-15-7 EINECS: 211-882-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...