3-Chlorbenzoylacetonitril
Lieferant: Thermo Scientific
Synonyme:
3-(3-Chlorphenyl)-3-oxopropionsäurenitril, 3-(3-Chlorphenyl)-3-ketopropionsäurenitril
KM01406DA
KM01406EA
MAYBKM01406DAEA
0
EUR
MAYBKM01406DA
MAYBKM01406EA
3-Chlorbenzoylacetonitril
3-Chlorbenzoylacetonitril
Formel:
C₉H₆ClNO Molecular Weight: 179,61 g/mol |
MDL:
MFCD00067891 CAS-Nummer: 21667-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...