2-Acetyl-4-bromthiophen

Lieferant: Thermo Scientific

KM10200DA KM10200EA
MAYBKM10200DAEA 0 EUR
MAYBKM10200DA MAYBKM10200EA
2-Acetyl-4-bromthiophen
2-Acetyl-4-bromthiophen
Formel: C₆H₅BrOS
Molecular Weight: 205.08 g/mol
MDL: MFCD01566552
CAS-Nummer: 7209-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR