3-Chlor-p-anisidin
Lieferant: CHEMPUR
Synonyme:
3-Chlor-4-anisidin, 3-Chlor-4-methoxyanilin, 4-Amino-2-chloranisol
NRB03411.250
NRB03411.1
CHMPNRB03411.250EA
0
EUR
CHMPNRB03411.250
MAYBNRB03411.1
3-Chlor-p-anisidin
3-Chlor-p-anisidin
Formel:
C₇H₈ClNO Molecular Weight: 157,6 g/mol Siedepunkt: 253 °C (1013 hPa) Schmelzpunkt: 82 °C |
MDL:
MFCD00007771 CAS-Nummer: 5345-54-0 EINECS: 226-298-4 UN: 2233 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...