3,5-Dimethyl-4-nitroisoxazol

Lieferant: Thermo Scientific

RF03204.1 RF03204.10 RF03204.50
MAYBRF03204.1EA 0 EUR
MAYBRF03204.1 MAYBRF03204.10 MAYBRF03204.50
3,5-Dimethyl-4-nitroisoxazol
3,5-Dimethyl-4-nitroisoxazole
Formel: C₅H₆N₂O₃
Molecular Weight: 142,11 g/mol
MDL: MFCD00051656
CAS-Nummer: 1123-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR