2-Amino-4,6-dimethylpyrimidin
Lieferant: CHEMPUR
Synonyme:
4,6-Dimethylpyrimidin-2-ylamin, 4,6-Dimethylpyrimidin-2-amin
RJC01263.250
RJC01263.1
CHMPRJC01263.250EA
0
EUR
CHMPRJC01263.250
MAYBRJC01263.1
2-Amino-4,6-dimethylpyrimidin
2-Amino-4,6-dimethylpyrimidine
Formel:
C₆H₉N₃ Molecular Weight: 123,16 g/mol Schmelzpunkt: 152…155 °C |
MDL:
MFCD00006102 CAS-Nummer: 767-15-7 EINECS: 212-181-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...