2'-Hydroxy-6'-methoxyacetophenon

Lieferant: CHEMPUR

RJC03747.250 RJC03747.1
CHMPRJC03747.250EA 0 EUR
CHMPRJC03747.250 MAYBRJC03747.1
2'-Hydroxy-6'-methoxyacetophenon
2'-Hydroxy-6'-methoxyacetophenon
Formel: C₉H₁₀O₃
MDL: MFCD00008732
CAS-Nummer: 703-23-1
EINECS: 211-872-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR