2,4-Dimethoxybenzylalkohol

Lieferant: CHEMPUR

RJC04015.250 RJC04015.1
CHMPRJC04015.250EA 0 EUR
CHMPRJC04015.250 MAYBRJC04015.1
2,4-Dimethoxybenzylalkohol
2,4-Dimethoxybenzylalkohol
Formel: C₉H₁₂O₃
Molecular Weight: 168.19 g/mol
Siedepunkt: 317
Schmelzpunkt: 34…35 °C
MDL: MFCD00004614
CAS-Nummer: 7314-44-5
EINECS: 230-775-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR