(Methoxymethyl)triphenylphosphoniumchlorid

Lieferant: CHEMPUR
Achtung

SB00615.250 SB00615.1
CHMPSB00615.250EA 0 EUR
CHMPSB00615.250 MAYBSB00615.1
(Methoxymethyl)triphenylphosphoniumchlorid
(Methoxymethyl)triphenylphosphonium chloride
Formel: C₂₀H₂₀ClOP
Molecular Weight: 342,8 g/mol
Schmelzpunkt: 190 °C
MDL: MFCD00011800
CAS-Nummer: 4009-98-7
EINECS: 223-664-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR