2-Isopropoxyphenol
Lieferant: CHEMPUR
SB00770.250
SB00770.1
CHMPSB00770.250EA
0
EUR
CHMPSB00770.250
MAYBSB00770.1
2-Isopropoxyphenol
2-Isopropoxyphenol
Formel:
C₉H₁₂O₂ Molecular Weight: 152,19 g/mol Siedepunkt: 63…66 °C (0,15 mmHg) Schmelzpunkt: 206…207 °C Dichte: 1,03 g/cm³ (20 °C) |
MDL:
MFCD00002184 CAS-Nummer: 4812-20-8 EINECS: 225-379-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...