2,6-Dichlorbenzothiazol
Lieferant: Thermo Scientific
SB01025DA
SB01025EA
MAYBSB01025DAEA
0
EUR
MAYBSB01025DA
MAYBSB01025EA
2,6-Dichlorbenzothiazol
2,6-Dichlorobenzothiazole
Formel:
C₇H₃Cl₂NS Molecular Weight: 204,08 g/mol |
MDL:
MFCD00044101 CAS-Nummer: 3622-23-9 EINECS: 222-819-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...