Isopropylcyanacetat

Lieferant: Thermo Scientific

Synonyme: Cyanessigsäureisopropylester

SB01086.1 SB01086.10 SB01086.50
MAYBSB01086.1EA 0 EUR
MAYBSB01086.1 MAYBSB01086.10 MAYBSB01086.50
Isopropylcyanacetat
Isopropylcyanacetat
Formel: C₆H₉NO₂
Molecular Weight: 127.14 g/mol
MDL: MFCD00019842
CAS-Nummer: 13361-30-3
EINECS: 236-421-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR