4-Isopropylphenylisothiocyanat

Lieferant: Thermo Scientific

SB01635DA SB01635EA SB01635.50
MAYBSB01635DAEA 0 EUR
MAYBSB01635DA MAYBSB01635EA MAYBSB01635.50
4-Isopropylphenylisothiocyanat
4-Isopropylphenylisothiocyanat
Formel: C₁₀H₁₁NS
Molecular Weight: 177,27 g/mol
Siedepunkt: 252 °C (1013 hPa)
Dichte: 1,03 g/cm³ (20 °C)
MDL: MFCD00022060
CAS-Nummer: 89007-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR