4-Ethoxyphenylisothiocyanat

Lieferant: Thermo Scientific

Synonyme: 1-Ethoxy-4-isothiocyanato-benzene, 4-Ethoxyphenylisothiocyanate

SB01663.1 SB01663EA SB01663.50
MAYBSB01663.1EA 0 EUR
MAYBSB01663.1 MAYBSB01663EA MAYBSB01663.50
4-Ethoxyphenylisothiocyanat
4-Ethoxyphenylisothiocyanat
Formel: C₉H₉NOS
Molecular Weight: 179.24 g/mol
Siedepunkt: 146 °C (19 mmHg)
Schmelzpunkt: 75…76 °C
MDL: MFCD00041096
CAS-Nummer: 3460-49-9
EINECS: 222-410-0
UN: 2923
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR