4-Methyl-2-nitrophenylisothiocyanat
Lieferant: Thermo Scientific
SB01776.1
SB01776.10
MAYBSB01776.1EA
0
EUR
MAYBSB01776.1
MAYBSB01776.10
4-Methyl-2-nitrophenylisothiocyanat
4-Methyl-2-nitrophenylisothiocyanat
Formel:
C₈H₆N₂O₂S Molecular Weight: 194,21 g/mol Schmelzpunkt: 67 °C |
MDL:
MFCD00041248 CAS-Nummer: 17614-74-3 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...