3-Chlor-2-methylphenylisocyanat

Lieferant: Thermo Scientific

Synonyme: Isocyansäure(3-chlor-2-methylphenyl)ester

SB01803.1 SB01803.10 SB01803.50
MAYBSB01803.1EA 0 EUR
MAYBSB01803.1 MAYBSB01803.10 MAYBSB01803.50
3-Chlor-2-methylphenylisocyanat
3-Chlor-2-methylphenylisocyanat
Formel: C₈H₆ClNO
Molecular Weight: 167,59 g/mol
Siedepunkt: 78…80 °C (1013 hPa)
Dichte: 1,247 g/cm³ (20 °C)
MDL: MFCD00037036
CAS-Nummer: 40397-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR