1-Amino-2-ethylbutan

Lieferant: Thermo Scientific

Synonyme: 2-Ethylbutylamin

SB01831.1 SB01831.10 SB01831.50
MAYBSB01831.1EA 0 EUR
MAYBSB01831.1 MAYBSB01831.10 MAYBSB01831.50
1-Amino-2-ethylbutan
1-Amino-2-ethylbutane
Formel: C₆H₁₅N
Molecular Weight: 101,19 g/mol
MDL: MFCD00040755
CAS-Nummer: 617-79-8
EINECS: 210-530-6
UN: 3286
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR