4-Chlor-3-nitrophenylisocyanat

Lieferant: Thermo Scientific

SB01901.1 SB01901.10 SB01901.50
MAYBSB01901.1EA 0 EUR
MAYBSB01901.1 MAYBSB01901.10 MAYBSB01901.50
4-Chlor-3-nitrophenylisocyanat
4-Chlor-3-nitrophenylisocyanat
Formel: C₇H₃ClN₂O₃
Molecular Weight: 198,57 g/mol
Schmelzpunkt: 65…67 °C
MDL: MFCD00037062
CAS-Nummer: 40397-96-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR