3-Chlor-4-fluorphenylisocyanat

Lieferant: CHEMPUR

Synonyme: 2-Chloro-1-fluoro-4-isocyanatobenzene

SB01907.250 SB01907.1
CHMPSB01907.250EA 0 EUR
CHMPSB01907.250 MAYBSB01907.1
3-Chlor-4-fluorphenylisocyanat
3-Chlor-4-fluorphenylisocyanat
Formel: C₇H₃ClFNO
Molecular Weight: 171,56 g/mol
Siedepunkt: 108…112 °C (1013 hPa)
Dichte: 1,377 g/cm³ (20 °C)
MDL: MFCD00037037
CAS-Nummer: 50529-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR