ω-Brom-3-chloracetophenon

Lieferant: Thermo Scientific

Synonyme: 2-Brom-3'-chloracetophenon, 3-Chlorphenacylbromid, α-Brom-3-chloracetophenon, Brommethyl-m-chlorphenylketon

SP00224 SP00224.0010
MAYBSP00224EA 0 EUR
MAYBSP00224 MAYBSP00224.0010
ω-Brom-3-chloracetophenon
ω-Brom-3-chloracetophenon
Formel: C₈H₆BrClO
Molecular Weight: 233,49 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00792669
CAS-Nummer: 41011-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR