α,3,5-Trichlortoluol

Lieferant: Thermo Scientific

Synonyme: 1,3-Dichlor-5-(chlormethyl)benzol, 3,5-Dichlorbenzylchlorid, 1-(Chlormethyl)-3,5-dichlorbenzol

SPB00444DA SPB00444EA
MAYBSPB00444DAEA 0 EUR
MAYBSPB00444DA MAYBSPB00444EA
α,3,5-Trichlortoluol
α,3,5-Trichlortoluol
Formel: C₇H₅Cl₃
Molecular Weight: 195,47 g/mol
MDL: MFCD00060278
CAS-Nummer: 3290-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR