2-Methoxy-5-(trifluormethyl)pyridin
Lieferant: Thermo Scientific
SPB05581.1
SPB05581.10
SPB05581.50
MAYBSPB05581.1EA
0
EUR
MAYBSPB05581.1
MAYBSPB05581.10
MAYBSPB05581.50
2-Methoxy-5-(trifluormethyl)pyridin
2-Methoxy-5-(trifluoromethyl)pyridine
Formel:
C₇H₆F₃NO |
MDL:
MFCD00153203 CAS-Nummer: 175277-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...