4-(Trifluormethyl)pyridin
Lieferant: CHEMPUR
SPB05714.250
SPB05714.1
CHMPSPB05714.250EA
0
EUR
CHMPSPB05714.250
MAYBSPB05714.1
4-(Trifluormethyl)pyridin
4-(Trifluoromethyl)pyridine
Formel:
C₆H₄F₃N Molecular Weight: 147,1 g/mol Siedepunkt: 105…106 °C (1013 hPa) |
MDL:
MFCD00153289 CAS-Nummer: 3796-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...