1-(2-Methylphenyl)-2-thioharnstoff
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methylphenylthioharnstoff, 1-(2-Tolyl)thioharnstoff, o-Tolylthioharnstoff, N-(2-Methylphenyl)thioharnstoff
SPB06632.10
SPB06632.50
MAYBSPB06632.10EA
0
EUR
MAYBSPB06632.10
MAYBSPB06632.50
1-(2-Methylphenyl)-2-thioharnstoff
1-(2-Methylphenyl)-2-thioharnstoff
Formel:
(C₆H₄(CH₃))NC(S)NH₂ Molecular Weight: 166.25 g/mol Siedepunkt: 295 Schmelzpunkt: 160 |
MDL:
MFCD00041162 CAS-Nummer: 614-78-8 EINECS: 210-395-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...