4-(Trifluormethyl)benzylamin

Lieferant: CHEMPUR

TL00150.250 TL00150.1
CHMPTL00150.250EA 0 EUR
CHMPTL00150.250 MAYBTL00150.1
4-(Trifluormethyl)benzylamin
4-(Trifluormethyl)benzylamin
Formel: C₈H₈F₃N
Molecular Weight: 175,15 g/mol
Siedepunkt: 95…97 °C (1013 hPa)
Dichte: 1,229 g/cm³ (20 °C)
MDL: MFCD00010220
CAS-Nummer: 3300-51-4
EINECS: 221-971-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR