N-Methyl-4-(trifluormethoxy)anilin

Lieferant: Thermo Scientific

TL00405DA TL00405EA
MAYBTL00405DAEA 0 EUR
MAYBTL00405DA MAYBTL00405EA
N-Methyl-4-(trifluormethoxy)anilin
N-Methyl-4-(trifluormethoxy)anilin
Formel: C₈H₈F₃NO
Molecular Weight: 191.15 g/mol
MDL: MFCD00052332
CAS-Nummer: 41419-59-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR