3,5-Dimethylisoxazol
Lieferant: Thermo Scientific
TL00573.1
TL00573EA
TL00573.50
MAYBTL00573.1EA
0
EUR
MAYBTL00573.1
MAYBTL00573EA
MAYBTL00573.50
3,5-Dimethylisoxazol
3,5-Dimethylisoxazole
Formel:
C₅H₇NO Molecular Weight: 97,12 g/mol Siedepunkt: 142 °C (1013 hPa) Dichte: 0,98 g/cm³ (20 °C) |
MDL:
MFCD00003156 CAS-Nummer: 300-87-8 EINECS: 206-100-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...